siRNA Details
| Gene Symbol: Rragc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017475.2 | NP_059503.2 | 5 | 933 | ||
| GenBank mRNA | AB017616.1 | - | 5 | 856 | ||
| AK150792.1 | - | 5 | 888 | |||
| AK159355.1 | - | 5 | 866 | |||
| AK159434.1 | - | 5 | 888 | |||
| AK166658.1 | - | 5 | 879 | |||
| BC005417.1 | - | 5 | 881 | 3495022 | ||
| BC037732.1 | - | 2 | 230 | |||
| BC071245.1 | - | 5 | 1281 | |||

Rragc
Genomic Map