siRNA Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017461.2 | NP_059489.2 | 9, 10 | 953 | ||
| GenBank mRNA | AV507599.1 | - | 1, 2 | 86 | ||
| AY974598.1 | - | 1, 2 | 73 | |||
| BC140998.1 | - | 9, 10 | 954 | 9055610 | ||
| BC145081.1 | - | 9, 10 | 954 | 9053629 | ||
| BX523183.1 | - | 2, 3 | 269 | |||
| M37030.1 | - | 9, 10 | 957 | |||

Sept1
Genomic Map