siRNA Details
| Gene Symbol: Atf7ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019426.2 | NP_062299.2 | 2 | 766 | ||
| GenBank mRNA | AJ132702.1 | - | 2 | 759 | ||
| AK082697.1 | - | 2 | 816 | |||
| AK134252.1 | - | 2 | 773 | |||
| AK167542.1 | - | 3 | 812 | |||
| BC022714.1 | - | 2 | 759 | |||
| BC037617.1 | - | 768 | ||||
| BC048827.1 | - | 3 | 927 | |||
| BC052460.1 | - | 2 | 766 | 6834550 | ||
| BC060246.1 | - | 2 | 749 | 5705945 | ||

Atf7ip
Genomic Map