siRNA Details
| Gene Symbol: Hils1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018792.1 | NP_061262.1 | 1 | 249 | ||
| GenBank mRNA | AB022320.1 | - | 1 | 249 | ||
| AB045724.1 | - | 1 | 242 | |||
| AK005718.1 | - | 1 | 280 | |||
| AY286319.1 | - | 1 | 249 | |||
| BC050966.1 | - | 1 | 282 | |||
| BC061119.1 | - | 1 | 312 | 6773493 | ||

Hils1
Genomic Map