siRNA Details
| Gene Symbol: Ndufa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019443.2 | NP_062316.1 | 1 | 168 | ||
| GenBank mRNA | AK007840.1 | - | 1 | 75 | ||
| AK131584.1 | - | 367 | ||||
| BC024364.1 | - | 1 | 77 | 5031206 | ||
| CT010398.1 | - | 1 | 20 | |||
| Y07708.1 | - | 1 | 187 | |||

Ndufa1
Genomic Map