siRNA Details
| Gene Symbol: Ndufa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019443.2 | NP_062316.1 | 2 | 287 | ||
| GenBank mRNA | AI844651.1 | - | 2 | 209 | ||
| AK007840.1 | - | 2 | 194 | |||
| AK131584.1 | - | 486 | ||||
| BC024364.1 | - | 2 | 196 | 5031206 | ||
| CF383819.1 | - | 1 | 71 | |||
| CT010398.1 | - | 2 | 139 | |||
| Y07708.1 | - | 2 | 306 | |||

Ndufa1
Genomic Map