siRNA Details
| Gene Symbol: Tbc1d32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033385.3 | NP_001028557.2 | 18 | 2089 | ||
| GenBank mRNA | AK141206.1 | - | 3 | 284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d32| Gene Symbol: Tbc1d32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033385.3 | NP_001028557.2 | 18 | 2089 | ||
| GenBank mRNA | AK141206.1 | - | 3 | 284 | ||