siRNA Details
| Gene Symbol: Arhgap27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205236.1 | NP_001192165.1 | 5 | 1709 | ||
| NM_133715.5 | NP_598476.2 | 3 | 678 | |||
| GenBank mRNA | AK046763.1 | - | 3 | 679 | ||
| AK143916.1 | - | 1 | 150 | |||
| AK154926.1 | - | 5 | 1710 | |||

Arhgap27
Genomic Map