siRNA Details
| Gene Symbol: Zkscan4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039115.2 | NP_001034204.1 | 2 | 757 | ||
| GenBank mRNA | BC141222.1 | - | 2 | 632 | 9055834 | |
| BC145543.1 | - | 2 | 545 | |||
| BU148250.1 | - | 2 | 699 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zkscan4| Gene Symbol: Zkscan4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039115.2 | NP_001034204.1 | 2 | 757 | ||
| GenBank mRNA | BC141222.1 | - | 2 | 632 | 9055834 | |
| BC145543.1 | - | 2 | 545 | |||
| BU148250.1 | - | 2 | 699 | |||