siRNA Details
Gene Symbol: Gm5800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034102.2 | NP_001029274.1 | 3 | 291 | ||
GenBank mRNA | BC049651.1 | - | 3 | 291 | ||
BC100479.1 | - | 3 | 291 | 30917737 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034102.2 | NP_001029274.1 | 3 | 291 | ||
GenBank mRNA | BC049651.1 | - | 3 | 291 | ||
BC100479.1 | - | 3 | 291 | 30917737 |