siRNA Details
| Gene Symbol: Wdr70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081402.1 | NP_001074871.1 | 10, 11 | 1176 | ||
| GenBank mRNA | AK159714.1 | - | 10, 11 | 1158 | ||
| AK160933.1 | - | 10, 11 | 1112 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr70| Gene Symbol: Wdr70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081402.1 | NP_001074871.1 | 10, 11 | 1176 | ||
| GenBank mRNA | AK159714.1 | - | 10, 11 | 1158 | ||
| AK160933.1 | - | 10, 11 | 1112 | |||