siRNA Details
| Gene Symbol: Kalrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164268.1 | NP_001157740.1 | 5 | 835 | ||
| NM_177357.3 | NP_796331.2 | 5 | 835 | |||
| GenBank mRNA | AK147314.1 | - | 5 | 857 | ||
| AK147518.1 | - | 5 | 857 | |||
| BC157950.1 | - | 5 | 835 | 9088125 | ||
| BC172101.1 | - | 5 | 835 | 9054387 | ||

Kalrn
Genomic Map