siRNA Details
| Gene Symbol: Baiap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163270.1 | NP_001156742.1 | 23 | 2284 | ||
| GenBank mRNA | AK122358.1 | - | 21 | 2403 | ||
| BC158116.1 | - | 23 | 2284 | 9088189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Baiap3| Gene Symbol: Baiap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163270.1 | NP_001156742.1 | 23 | 2284 | ||
| GenBank mRNA | AK122358.1 | - | 21 | 2403 | ||
| BC158116.1 | - | 23 | 2284 | 9088189 | ||