siRNA Details
| Gene Symbol: Hmcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024720.3 | NP_001019891.2 | 72 | 11496 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmcn1| Gene Symbol: Hmcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024720.3 | NP_001019891.2 | 72 | 11496 | ||