siRNA Details
Gene Symbol: Cep170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099637.2 | NP_001093107.2 | 20 | 4633 | ||
GenBank mRNA | AK016937.1 | - | 8 | 894 | ||
AK038878.1 | - | 7 | 1202 | |||
AK078541.1 | - | 3 | 115 | |||
AK172953.1 | - | 8 | 2142 | |||
BC038377.1 | - | 8 | 2094 | |||
BC085106.1 | - | 7 | 1741 |
