siRNA Details
| Gene Symbol: Cep170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099637.2 | NP_001093107.2 | 5 | 475 | ||
| GenBank mRNA | BC054781.1 | - | 5 | 521 | ||
| BC057019.1 | - | 5 | 521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep170| Gene Symbol: Cep170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099637.2 | NP_001093107.2 | 5 | 475 | ||
| GenBank mRNA | BC054781.1 | - | 5 | 521 | ||
| BC057019.1 | - | 5 | 521 | |||