siRNA Details
| Gene Symbol: Erich6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081262.1 | NP_001074731.1 | 13 | 1744 | ||
| GenBank mRNA | AK133299.1 | - | 10 | 2346 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich6| Gene Symbol: Erich6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081262.1 | NP_001074731.1 | 13 | 1744 | ||
| GenBank mRNA | AK133299.1 | - | 10 | 2346 | ||