siRNA Details
| Gene Symbol: Nup210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018815.2 | NP_061285.2 | 15 | 2112 | ||
| GenBank mRNA | AF113751.1 | - | 15 | 2080 | ||
| AK157272.1 | - | 15 | 2104 | |||
| AK173056.1 | - | 15 | 2066 | |||
| BC052468.1 | - | 15 | 2105 | 6835038 | ||

Nup210
Genomic Map