siRNA Details
| Gene Symbol: Zar1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159693.1 | NP_001153165.1 | 1 | 442 | ||
| GenBank mRNA | FJ858201.1 | - | 1 | 442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zar1l| Gene Symbol: Zar1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159693.1 | NP_001153165.1 | 1 | 442 | ||
| GenBank mRNA | FJ858201.1 | - | 1 | 442 | ||