siRNA Details
| Gene Symbol: Ptprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207270.2 | NP_997153.2 | 2, 3 | 425 | ||
| GenBank mRNA | AB217856.1 | - | 1, 2 | 98 | ||
| AK078884.1 | - | 1, 2 | 99 | |||
| BB618721.1 | - | 2, 3 | 425 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprh| Gene Symbol: Ptprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207270.2 | NP_997153.2 | 2, 3 | 425 | ||
| GenBank mRNA | AB217856.1 | - | 1, 2 | 98 | ||
| AK078884.1 | - | 1, 2 | 99 | |||
| BB618721.1 | - | 2, 3 | 425 | |||