siRNA Details
| Gene Symbol: Gm5897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136397.1 | - | 1156 | |||
| BC147460.1 | - | 1 | 681 | 9056102 | ||
| BC147474.1 | - | 1 | 681 | 9056116 | ||
| Gene Symbol: Lmtk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029204.1 | - | 708 | |||

Gm5897
Genomic Map