siRNA Details
| Gene Symbol: Fam160a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242363.2 | NP_001229292.1 | 13 | 7003 | ||
| NM_001242364.2 | NP_001229293.1 | 12 | 6954 | |||
| NM_001242365.2 | NP_001229294.1 | 12 | 6360 | |||
| NM_199009.3 | NP_950174.1 | 12 | 6912 | |||
| Gene Symbol: Gm5901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195727.1 | NP_001182656.1 | 1 | 51 | ||

Fam160a2
Genomic Map