siRNA Details
| Gene Symbol: Gm5901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195727.1 | NP_001182656.1 | 2 | 565 | ||
| Gene Symbol: Fam160a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242363.2 | NP_001229292.1 | 13 | 4560 | ||
| NM_001242364.2 | NP_001229293.1 | 12 | 4511 | |||
| NM_001242365.2 | NP_001229294.1 | 12 | 3917 | |||
| NM_199009.3 | NP_950174.1 | 12 | 4469 | |||
| GenBank mRNA | AK014591.1 | - | 1 | 1351 | ||
| AK122552.1 | - | 11 | 4218 | |||

Gm5901
Genomic Map