siRNA Details
Gene Symbol: 7420426K07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033983.3 | NP_001029155.1 | 1 | 228 | ||
GenBank mRNA | AK135855.1 | - | 1 | 229 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 7420426K07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033983.3 | NP_001029155.1 | 1 | 228 | ||
GenBank mRNA | AK135855.1 | - | 1 | 229 |