siRNA Details
Gene Symbol: Wdr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290792.1 | NP_001277721.1 | 10 | 1080 | ||
NM_001290794.1 | NP_001277723.1 | 10 | 976 | |||
NM_001290795.1 | NP_001277724.1 | 10 | 1038 | |||
NM_172372.2 | NP_758960.1 | 10 | 1076 | |||
GenBank mRNA | AF229635.1 | - | 10 | 1012 | ||
AK008427.1 | - | 2 | 125 | |||
AK018409.1 | - | 10 | 1076 | |||
AK145616.1 | - | 10 | 976 | |||
BC011479.1 | - | 10 | 1047 | 4035468 |
