siRNA Details
Gene Symbol: Wdr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290792.1 | NP_001277721.1 | 11 | 1220 | ||
NM_001290794.1 | NP_001277723.1 | 11 | 1116 | |||
NM_001290795.1 | NP_001277724.1 | 11 | 1178 | |||
NM_172372.2 | NP_758960.1 | 11 | 1216 | |||
GenBank mRNA | AF229635.1 | - | 11 | 1152 | ||
AI854309.1 | - | 1 | 393 | |||
AK008427.1 | - | 3 | 265 | |||
AK018409.1 | - | 11 | 1216 | |||
AK145616.1 | - | 11 | 1116 | |||
BC011479.1 | - | 11 | 1187 | 4035468 |
