siRNA Details
| Gene Symbol: Sh3glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282037.1 | NP_001268966.1 | 11 | 1425 | ||
| NM_019464.3 | NP_062337.1 | 9 | 1362 | |||
| GenBank mRNA | AF263364.1 | - | 9 | 1350 | ||
| AF272946.1 | - | 11 | 1413 | |||
| AK004590.1 | - | 9 | 1173 | |||
| AK029647.1 | - | 8 | 1023 | |||
| AK146246.1 | - | 9 | 1189 | |||
| AK172959.1 | - | 9 | 1173 | |||
| BC024362.1 | - | 9 | 1172 | 5031296 | ||

Sh3glb1
Genomic Map