siRNA Details
| Gene Symbol: Plxnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018797.2 | NP_061267.1 | 22 | 3662 | ||
| GenBank mRNA | AF190578.1 | - | 22 | 3662 | ||
| AK033998.1 | - | 4 | 522 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plxnc1| Gene Symbol: Plxnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018797.2 | NP_061267.1 | 22 | 3662 | ||
| GenBank mRNA | AF190578.1 | - | 22 | 3662 | ||
| AK033998.1 | - | 4 | 522 | |||