siRNA Details
| Gene Symbol: Parp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039530.3 | NP_001034619.2 | 6 | 2037 | ||
| GenBank mRNA | AK089685.1 | - | 7 | 2128 | ||
| AK156593.1 | - | 5 | 3703 | |||
| BC138874.1 | - | 6 | 2152 | 8861896 | ||
| BC145452.1 | - | 6 | 2152 | |||
| DQ372930.1 | - | 6 | 2201 | |||

Parp14
Genomic Map