siRNA Details
| Gene Symbol: Ugt2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024148.1 | NP_001019319.1 | 2 | 93 | ||
| GenBank mRNA | BC048920.1 | - | 1 | 94 | ||
| BC058786.1 | - | 2 | 93 | 6310060 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt2a2| Gene Symbol: Ugt2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024148.1 | NP_001019319.1 | 2 | 93 | ||
| GenBank mRNA | BC048920.1 | - | 1 | 94 | ||
| BC058786.1 | - | 2 | 93 | 6310060 | ||