siRNA Details
| Gene Symbol: Ctps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168568.1 | NP_001162040.1 | 12, 13 | 1476 | ||
| NM_001168569.1 | NP_001162041.1 | 12, 13 | 1533 | |||
| NM_001168571.1 | NP_001162043.1 | 12, 13 | 1608 | |||
| NM_018737.5 | NP_061207.1 | 12, 13 | 1608 | |||
| GenBank mRNA | AK032018.1 | - | 12, 13 | 1335 | ||
| AK052854.1 | - | 12, 13 | 1524 | |||
| AK084595.1 | - | 12, 13 | 1405 | |||
| AK150966.1 | - | 12, 13 | 1475 | |||
| AK151597.1 | - | 12, 13 | 1475 | |||
| AK161564.1 | - | 12, 13 | 1446 | |||
| AK164206.1 | - | 12, 13 | 1435 | |||
| BC003257.1 | - | 12, 13 | 1316 | 3500778 | ||
| U49385.1 | - | 12, 13 | 1498 | |||

Ctps2
Genomic Map