siRNA Details
Gene Symbol: Stx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018768.2 | NP_061238.1 | 5 | 414 | ||
GenBank mRNA | AB040054.1 | - | 5 | 411 | ||
AF036716.1 | - | 5 | 401 | |||
AF093064.1 | - | 5 | 423 | |||
AK003297.1 | - | 5 | 440 | |||
AK018716.1 | - | 4 | 241 | |||
BC038186.1 | - | 5 | 413 | |||
BC048167.1 | - | 5 | 414 | 6821351 | ||
BC048479.1 | - | 5 | 439 | 6774827 |
