siRNA Details
| Gene Symbol: Stx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018768.2 | NP_061238.1 | 6 | 482 | ||
| GenBank mRNA | AB040054.1 | - | 6 | 479 | ||
| AF036716.1 | - | 6 | 469 | |||
| AF093064.1 | - | 6 | 491 | |||
| AK003297.1 | - | 6 | 508 | |||
| AK018716.1 | - | 5 | 309 | |||
| BC038186.1 | - | 6 | 481 | |||
| BC048167.1 | - | 6 | 482 | 6821351 | ||
| BC048479.1 | - | 6 | 507 | 6774827 | ||

Stx8
Genomic Map