siRNA Details
| Gene Symbol: Stx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018768.2 | NP_061238.1 | 7, 8 | 650 | ||
| GenBank mRNA | AB040054.1 | - | 7, 8 | 647 | ||
| AF036716.1 | - | 7, 8 | 637 | |||
| AF093064.1 | - | 7, 8 | 659 | |||
| AK003297.1 | - | 7, 8 | 676 | |||
| AK018716.1 | - | 6, 7 | 477 | |||
| BC038186.1 | - | 7, 8 | 649 | |||
| BC048167.1 | - | 7, 8 | 650 | 6821351 | ||
| BC048479.1 | - | 7, 8 | 675 | 6774827 | ||

Stx8
Genomic Map