siRNA Details
| Gene Symbol: Sfn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018754.2 | NP_061224.2 | 1 | 522 | ||
| GenBank mRNA | AF058798.1 | - | 1 | 286 | ||
| AF152893.1 | - | 1 | 123 | |||
| AK146490.1 | - | 1 | 196 | |||
| AK169358.1 | - | 1 | 187 | |||
| CX224942.1 | - | 1 | 585 | |||

Sfn
Genomic Map