siRNA Details
| Gene Symbol: Sfn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018754.2 | NP_061224.2 | 1 | 832 | ||
| GenBank mRNA | AF058798.1 | - | 1 | 596 | ||
| AF152893.1 | - | 1 | 433 | |||
| AK146490.1 | - | 1 | 506 | |||
| AK169358.1 | - | 1 | 497 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sfn| Gene Symbol: Sfn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018754.2 | NP_061224.2 | 1 | 832 | ||
| GenBank mRNA | AF058798.1 | - | 1 | 596 | ||
| AF152893.1 | - | 1 | 433 | |||
| AK146490.1 | - | 1 | 506 | |||
| AK169358.1 | - | 1 | 497 | |||