siRNA Details
| Gene Symbol: Olfr70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019485.2 | NP_062358.2 | 2 | 818 | ||
| GenBank mRNA | BC046311.1 | - | 2 | 818 | 6307520 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr70| Gene Symbol: Olfr70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019485.2 | NP_062358.2 | 2 | 818 | ||
| GenBank mRNA | BC046311.1 | - | 2 | 818 | 6307520 | |