siRNA Details
| Gene Symbol: Plac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019538.4 | NP_062411.1 | 3 | 438 | ||
| GenBank mRNA | AF234653.1 | - | 3 | 439 | ||
| AF250838.1 | - | 3 | 435 | |||
| AK005501.1 | - | 3 | 439 | |||
| AK145876.1 | - | 3 | 404 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plac1| Gene Symbol: Plac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019538.4 | NP_062411.1 | 3 | 438 | ||
| GenBank mRNA | AF234653.1 | - | 3 | 439 | ||
| AF250838.1 | - | 3 | 435 | |||
| AK005501.1 | - | 3 | 439 | |||
| AK145876.1 | - | 3 | 404 | |||