siRNA Details
| Gene Symbol: Grasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019518.3 | NP_062391.3 | 7 | 740 | ||
| GenBank mRNA | AB041603.1 | - | 7 | 738 | ||
| AF236099.1 | - | 7 | 703 | |||
| BC046307.1 | - | 7 | 708 | 6306542 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grasp| Gene Symbol: Grasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019518.3 | NP_062391.3 | 7 | 740 | ||
| GenBank mRNA | AB041603.1 | - | 7 | 738 | ||
| AF236099.1 | - | 7 | 703 | |||
| BC046307.1 | - | 7 | 708 | 6306542 | ||