siRNA Details
| Gene Symbol: Bace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019517.4 | NP_062390.3 | 2 | 439 | ||
| GenBank mRNA | AF216310.1 | - | 2 | 390 | ||
| AK052309.1 | - | 2 | 426 | |||
| AK078770.1 | - | 2 | 406 | |||
| AK165036.1 | - | 2 | 375 | |||
| AK186833.1 | - | 1 | 73 | |||
| BC120773.1 | - | 2 | 405 | 40129696 | ||
| BC131947.1 | - | 2 | 405 | 40130224 | ||

Bace2
Genomic Map