siRNA Details
| Gene Symbol: Rbm38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019547.2 | NP_062420.2 | 2 | 397 | ||
| GenBank mRNA | AI573584.1 | - | 2 | 415 | ||
| AK146026.1 | - | 2 | 371 | |||
| AK162141.1 | - | 2 | 370 | |||
| AK166609.1 | - | 2 | 398 | |||
| AK172625.1 | - | 2 | 368 | |||
| BC006687.1 | - | 1 | 906 | |||
| BC085307.1 | - | 2 | 341 | 6396103 | ||

Rbm38
Genomic Map