siRNA Details
| Gene Symbol: Heyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013905.3 | NP_038933.2 | 5 | 652 | ||
| GenBank mRNA | AB093590.1 | - | 5 | 344 | ||
| AF172288.1 | - | 5 | 652 | |||
| AJ271868.2 | - | 5 | 652 | |||
| AK004697.2 | - | 5 | 355 | |||
| AK028596.1 | - | 5 | 391 | |||
| AK080822.1 | - | 5 | 356 | |||
| BC130263.1 | - | 5 | 360 | 40142873 | ||

Heyl
Genomic Map