siRNA Details
| Gene Symbol: Cpxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019696.2 | NP_062670.2 | 11 | 1696 | ||
| GenBank mRNA | AF077738.1 | - | 11 | 1641 | ||
| AK132611.1 | - | 7 | 942 | |||
| BC003713.1 | - | 11 | 1629 | 3593211 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpxm1| Gene Symbol: Cpxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019696.2 | NP_062670.2 | 11 | 1696 | ||
| GenBank mRNA | AF077738.1 | - | 11 | 1641 | ||
| AK132611.1 | - | 7 | 942 | |||
| BC003713.1 | - | 11 | 1629 | 3593211 | ||