siRNA Details
| Gene Symbol: Stk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019635.2 | NP_062609.2 | 11 | 1730 | ||
| GenBank mRNA | AF271361.1 | - | 11 | 1476 | ||
| AK002914.1 | - | 6 | 948 | |||
| AK004141.1 | - | 2 | 306 | |||
| AY058922.1 | - | 11 | 1616 | |||
| BC037440.1 | - | 9 | 1486 | 3585312 | ||
| BC049123.1 | - | 11 | 1604 | 6813956 | ||
| U28726.1 | - | 11 | 1608 | |||

Stk3
Genomic Map