siRNA Details
| Gene Symbol: Naa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177965.1 | NP_001171436.1 | 8 | 772 | ||
| NM_019870.3 | NP_063923.1 | 8 | 681 | |||
| GenBank mRNA | AJ627039.1 | - | 7 | 500 | ||
| AK003806.1 | - | 8 | 709 | |||
| AK009697.1 | - | 8 | 683 | |||
| AK014469.1 | - | 8 | 610 | |||
| AY183136.1 | - | 8 | 650 | |||
| AY183137.1 | - | 8 | 741 | |||
| BC027219.1 | - | 8 | 673 | 3673739 | ||

Naa10
Genomic Map