siRNA Details
| Gene Symbol: Slc35g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019871.2 | NP_063924.2 | 2 | 707 | ||
| GenBank mRNA | AF016712.1 | - | 2 | 557 | ||
| AK077181.1 | - | 2 | 833 | |||
| BC137893.1 | - | 2 | 587 | 8860912 | ||
| BC137894.1 | - | 2 | 587 | 8860913 | ||

Slc35g3
Genomic Map