siRNA Details
Gene Symbol: Fam60a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019643.3 | NP_062617.2 | 6 | 721 | ||
GenBank mRNA | AK046519.1 | - | 6 | 628 | ||
AK145384.1 | - | 6 | 732 | |||
AK166569.1 | - | 6 | 727 | |||
BC076599.1 | - | 6 | 721 | 30614283 | ||
U64033.1 | - | 6 | 810 |
