siRNA Details
| Gene Symbol: Gps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019726.3 | NP_062700.2 | 4 | 502 | ||
| GenBank mRNA | AF153696.1 | - | 5 | 442 | ||
| BC011317.1 | - | 4 | 502 | 3979292 | ||
| BC138879.1 | - | 4 | 458 | 8861901 | ||
| BC138880.1 | - | 4 | 459 | 8861902 | ||

Gps2
Genomic Map