siRNA Details
Gene Symbol: Zfp113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019747.4 | NP_062721.2 | 5 | 799 | ||
GenBank mRNA | AK076349.1 | - | 5 | 794 | ||
AK220556.1 | - | 5 | 799 | |||
BC052453.1 | - | 5 | 743 | 6410632 | ||
BC054525.1 | - | 5 | 743 | 6410632 | ||
BC056445.1 | - | 4 | 664 | 5703904 | ||
BC056991.1 | - | 5 | 745 |
