siRNA Details
| Gene Symbol: Aatf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019816.1 | NP_062790.1 | 12 | 1593 | ||
| GenBank mRNA | AF222801.1 | - | 12 | 1593 | ||
| AK016820.1 | - | 8 | 830 | |||
| AK141334.1 | - | 12 | 1591 | |||
| BC025080.1 | - | 12 | 1580 | 2648117 | ||

Aatf
Genomic Map